Learn More
Braintree Scientific Orbi-Shaker Jr. tube rack 12x50 & 21x15ml conical tubes max2
SDP
Catalog No. 501954450
Encompass
Perfectly circular 19mm orbit for aeration and mixing. Digital speed control up to 300 rpm. The Orbi-Shaker JR. is a space-saving shaker, complete with a 9.5" x 8" non slip, rubber coated platform that is ideal for tissue culture flasks, petri dishes and staining trays. Additional platforms are also available for the Orbi-Shaker JR, including the convenient MAGic Clamp™ platform (patent pending) for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes. Technical Data: Speed range 50 to 300 rpm, Speed increment: 10 rpm, Orbit: 19mm (3/4"), Maximum capacity: 4 x 1L or 4 kg, Standard platform dimensions: 9.5" x 8"/24cm x 20cm, Dimensions (WxDxH): 11" x 10.7" x 5"/28cm x 27cm x 13cm, Operating Temperature: 4 to 650C **Sold for research purposes only and not for human use**
Catalog No. 50-195-4450 Supplier Braintree Scientific Supplier No. BT302MR1550
Perfectly circular 19mm orbit for aeration and mixing. Digital speed control up to 300 rpm. The Orbi-Shaker JR. is a space-saving shaker, complete with a 9.5" x 8" non slip, rubber coated platform that is ideal for tissue culture flasks, petri dishes and staining trays. Additional platforms are also available for the Orbi-Shaker JR, including the convenient MAGic Clamp™ platform (patent pending) for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes. Technical Data: Speed range 50 to 300 rpm, Speed increment: 10 rpm, Orbit: 19mm (3/4"), Maximum capacity: 4 x 1L or 4 kg, Standard platform dimensions: 9.5" x 8"/24cm x 20cm, Dimensions (WxDxH): 11" x 10.7" x 5"/28cm x 27cm x 13cm, Operating Temperature: 4 to 650C **Sold for research purposes only and not for human use**