Learn More
Thomas Scientific Orbi-Shaker JR 9.5x8" platform US plug
SDP
Catalog No. 501461096
Encompass_Preferred
Compact and economically priced Perfect for low capacity applications Incubator and cold room safe The Orbi-Shaker JR.™ is a personal sized orbital shaker including a 9.5x8” non-slip rubber platform. The Orbi-Shaker Jr. is the ideal choice for mixing a small quantity of flasks or other vessels. The Orbi-Shaker JR. is a personal sized orbital shaker including a 9.5x8” non-slip rubber platform, ideal for tissue culture flasks, Petri dishes and staining trays. Additional platforms are also available for the Orbi-Shaker JR, including the convenient MAGic Clamp™ platform (PATENT PENDING) for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes. Mixing speed and run time are displayed on the large LED control panel located on the front side of the unit. The integral microprocessor includes a constant monitoring system that ensures accuracy through the durat
Catalog No. 50-146-1096 Supplier Thomas Scientific Supplier No. 1222U29
Compact and economically priced Perfect for low capacity applications Incubator and cold room safe The Orbi-Shaker JR.™ is a personal sized orbital shaker including a 9.5x8” non-slip rubber platform. The Orbi-Shaker Jr. is the ideal choice for mixing a small quantity of flasks or other vessels. The Orbi-Shaker JR. is a personal sized orbital shaker including a 9.5x8” non-slip rubber platform, ideal for tissue culture flasks, Petri dishes and staining trays. Additional platforms are also available for the Orbi-Shaker JR, including the convenient MAGic Clamp™ platform (PATENT PENDING) for use with Erlenmeyer flasks and test tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes. Mixing speed and run time are displayed on the large LED control panel located on the front side of the unit. The integral microprocessor includes a constant monitoring system that ensures accuracy through the durat