Learn More
Research Products International Corp Orbi-Shaker Jr., Compact Orbital Shaker with Flat Mat Platform
SDP
Catalog No. 50998290
Encompass
Convenient size orbital shaker fits easily on a benchtop or in cold room or incubator. Compact design measures 11" x 10 3/4" x 5"H and weighs only 12 pounds. Ideal for a variety of laboratory applications that requires unattended mixing.
Features include high tech microprocessor monitoring system that ensures maximum accuracy and safety through the entire cycle. Supplied with standard non-slip mat platform (9 1/2" x 8"), instructions and a two year warranty.
Catalog No. 50-998-290 Supplier Research Products International Corp Supplier No. 260140
Convenient size orbital shaker fits easily on a benchtop or in cold room or incubator. Compact design measures 11" x 10 3/4" x 5"H and weighs only 12 pounds. Ideal for a variety of laboratory applications that requires unattended mixing.
Features include high tech microprocessor monitoring system that ensures maximum accuracy and safety through the entire cycle. Supplied with standard non-slip mat platform (9 1/2" x 8"), instructions and a two year warranty.