Learn More
Research Products International Corp Orbi-Shaker XL
SDP
Catalog No. NC1634094
Encompass
The Orbi-Shaker XL offers new, full color TOUCH screen control panel and a flask capacity that far exceeds that of most benchtop models. With a large 18 x 18 inch platform and a powerful, beltless motor drive, the Orbi-Shaker XL is capable of mixing up to five 2L flasks or nine 1L flasks simultaneously. The included, non-slip rubber mat platform is ideal for low speed applications.
Catalog No. NC1634094 Supplier Research Products International Corp Supplier No. 260115
The Orbi-Shaker XL offers new, full color TOUCH screen control panel and a flask capacity that far exceeds that of most benchtop models. With a large 18 x 18 inch platform and a powerful, beltless motor drive, the Orbi-Shaker XL is capable of mixing up to five 2L flasks or nine 1L flasks simultaneously. The included, non-slip rubber mat platform is ideal for low speed applications.