Learn More
Sigma Aldrich Fine Chemicals Biosciences Orbi ShakerTM CO2 with rem1EA
Catalog No. 502908900
Encompass
The Orbi-Shaker CO2 is designed for shaking mixing and aeration applications in extreme environments including CO2 incubators. It can withstand temperatures from 0 to 60C and humidity levels up to 100%.To avoid contamination unintentional release of gas or deviations from protocol both speed and time are set via the included remote control module. The remote controller allows the user to set and adjust the shaker settings without opening the chamber door(s).The instrument is supplied with a 13 x 12 in. non-slip platform..nbspDesigned for use in C02 incubators other extreme environments.nbspRemote controlled adjust settings from outside the chamber.nbspHorizontally circular 19 mm orbit for aeration mixing.nbspInstantly exchange flask clamps with MAGic Clamp(TM) accessories
Catalog No. 50-290-8900 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7634381EA
The Orbi-Shaker CO2 is designed for shaking mixing and aeration applications in extreme environments including CO2 incubators. It can withstand temperatures from 0 to 60C and humidity levels up to 100%.To avoid contamination unintentional release of gas or deviations from protocol both speed and time are set via the included remote control module. The remote controller allows the user to set and adjust the shaker settings without opening the chamber door(s).The instrument is supplied with a 13 x 12 in. non-slip platform..nbspDesigned for use in C02 incubators other extreme environments.nbspRemote controlled adjust settings from outside the chamber.nbspHorizontally circular 19 mm orbit for aeration mixing.nbspInstantly exchange flask clamps with MAGic Clamp(TM) accessories