Learn More
Sigma Aldrich Fine Chemicals Biosciences Orbi ShakerTM CO2 XL orbit1EA
Catalog No. 502908634
Encompass
The Orbi-Shaker CO2 XL is designed for shaking mixing and aeration applications in extreme environments including CO2 incubators. It can withstand temperatures from 0 to 60C and humidity levels up to 100%. To avoid contamination unintentional release of gas or deviations from protocol all settings are adjusted via the included remote control module.This remote control module can be stored outside of the chamber and connects to the shaker via flat (1 mm thickness) cable that easily passes through a port or underneath an incubator door without compromising the seal. This allows for user adjustment of run time or mixing speed without opening the chamber door(s).The Orbi-Shaker CO2 XL is supplied with a 16 x 16.5 inch non-slip rubber mat platform.
Catalog No. 50-290-8634 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7413361EA
Add to Cart
Add to Cart
Request a Quote
The Orbi-Shaker CO2 XL is designed for shaking mixing and aeration applications in extreme environments including CO2 incubators. It can withstand temperatures from 0 to 60C and humidity levels up to 100%. To avoid contamination unintentional release of gas or deviations from protocol all settings are adjusted via the included remote control module.This remote control module can be stored outside of the chamber and connects to the shaker via flat (1 mm thickness) cable that easily passes through a port or underneath an incubator door without compromising the seal. This allows for user adjustment of run time or mixing speed without opening the chamber door(s).The Orbi-Shaker CO2 XL is supplied with a 16 x 16.5 inch non-slip rubber mat platform.