Learn More
Sigma Aldrich Fine Chemicals Biosciences Orbi ShakerTM MP with 4 po1EA
Catalog No. 502908897
Encompass
The Benchmark Scientific Orbi-Shaker MP is designed with a 3mm mixing orbit optimized for thorough mixing of microplates and/or PCR plates. Up to four plates (standard or deep well) can be placed on the included platform and are instantly secured without the use springs clamps or tools.A digital microprocessor ensures accurate speed control over the brushless maintenance free motor drive and both speed and time can be viewed simultaneously on the large LED display.
Catalog No. 50-290-8897 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7424501EA
The Benchmark Scientific Orbi-Shaker MP is designed with a 3mm mixing orbit optimized for thorough mixing of microplates and/or PCR plates. Up to four plates (standard or deep well) can be placed on the included platform and are instantly secured without the use springs clamps or tools.A digital microprocessor ensures accurate speed control over the brushless maintenance free motor drive and both speed and time can be viewed simultaneously on the large LED display.