Learn More
Sigma Aldrich Fine Chemicals Biosciences Orbi ShakerTM orbital shak1EA
Catalog No. 502909007
Encompass
The powerful Orbi-Shaker is capable of mixing a 14 lb load capacity up to speeds as high as 300 rpm. The standard platform (included) features a non-slip rubber coated surface ideal for tissue culture flasks petri dishes and staining trays.High capacity up to 9 x 500 mL 5 x 1 L or 2 x 2 LHorizontally circular 19 mm orbit for aeration mixingDigital speed control up to 300 rpmInstantly exchange flask clamps with MAGic Clamp(TM) accessories
Catalog No. 50-290-9007 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7654651EA
The powerful Orbi-Shaker is capable of mixing a 14 lb load capacity up to speeds as high as 300 rpm. The standard platform (included) features a non-slip rubber coated surface ideal for tissue culture flasks petri dishes and staining trays.High capacity up to 9 x 500 mL 5 x 1 L or 2 x 2 LHorizontally circular 19 mm orbit for aeration mixingDigital speed control up to 300 rpmInstantly exchange flask clamps with MAGic Clamp(TM) accessories