Learn More
Sigma Aldrich Fine Chemicals Biosciences Orbi ShakerTM XL orbital s1EA
Catalog No. 502908899
Encompass
The Orbi-Shaker XL offers flask capacity that far exceeds most bench-top models. With the large platform and powerful beltless motor drive the Orbi-Shaker XL is capable of mixing up to 5 x 2 L flasks or 9 x 1 L flasks simultaneously. The included non-slip rubber mat platform is ideal for low speed applications while the convenient MAGic Clamp(TM) platform is also available for high speed mixing of Erlenmeyer flasks tube racks or microplates. The MAGic Clamp(TM) allows for instant flask clamp exchange without the use of tools screws or any other hardware.
Catalog No. 50-290-8899 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7633731EA
The Orbi-Shaker XL offers flask capacity that far exceeds most bench-top models. With the large platform and powerful beltless motor drive the Orbi-Shaker XL is capable of mixing up to 5 x 2 L flasks or 9 x 1 L flasks simultaneously. The included non-slip rubber mat platform is ideal for low speed applications while the convenient MAGic Clamp(TM) platform is also available for high speed mixing of Erlenmeyer flasks tube racks or microplates. The MAGic Clamp(TM) allows for instant flask clamp exchange without the use of tools screws or any other hardware.