Learn More
Cayman Chemical Orbifloxacin, Purity: ≥98%, CAS: 113617-63-3, 50 mg
SDP
Catalog No. 502979430
Encompass_Preferred
A fluoroquinolone antibiotic used in animals, including dogs, cattle, and swine, to combat gastrointestinal and respiratory infections
Catalog No. 50-297-9430 Supplier Cayman Chemical Supplier No. 1735350 MG
A fluoroquinolone antibiotic used in animals, including dogs, cattle, and swine, to combat gastrointestinal and respiratory infections