Learn More
MSC Orbis Plastic Stack-N-Nest Container, Grey, 23.6 x 15.8 x 11.3 in
Catalog No. 5000533807
Encompass
This grey plastic storage container features an attached lid and is designed for efficient material handling. It is part of the 4048 Stack-N-Nest System, an integrated solution that allows these reusable plastic totes to stack in a column, creating vertically modular loads and consistent stack heights. It can also be made with recycled plastics.
  • 24 x 16 footprint
  • Attached lid design
  • Stacks with the lid closed and nests when empty
  • Solid walls
  • Smooth or textured bottom options available
  • Available in FDA-approved material
  • Available in ESD-protective material
  • Contoured handles for easy and safe handling
  • Identification areas on sides, ends, and covers
  • Fully recyclable at the end of service life
  • 3.0:1 nesting ratio
Catalog No. 50-005-33807 Supplier MSC Supplier No. 43399179
This grey plastic storage container features an attached lid and is designed for efficient material handling. It is part of the 4048 Stack-N-Nest System, an integrated solution that allows these reusable plastic totes to stack in a column, creating vertically modular loads and consistent stack heights. It can also be made with recycled plastics.
  • 24 x 16 footprint
  • Attached lid design
  • Stacks with the lid closed and nests when empty
  • Solid walls
  • Smooth or textured bottom options available
  • Available in FDA-approved material
  • Available in ESD-protective material
  • Contoured handles for easy and safe handling
  • Identification areas on sides, ends, and covers
  • Fully recyclable at the end of service life
  • 3.0:1 nesting ratio