Learn More
Chemglass Life Sciences OrbiShaker CO², Remote Controlled Orbital Shaker, with Microplate Platform
SDP
Supplier:  Chemglass Life Sciences CLS2450M01
Encompass
Designed for use in CO2 Incubators and other extreme environments. Remote control, adjust settings without exposing chamber.compact, 3mm orbit for thorough mixing of well plates. Includes 6-position microplate platform. TheOrbi-Shaker CO2-MP is designed for thorough, mixing of microplates in extreme environments, including CO2 incubators. It can withstand temperatures from 0 to 65 degreesC and humidity levels up to 100%. To avoid contamination, unintentional release of gas or deviations from protocol, both speed and time are set via the included remote-control module. The remote controller can magnetically mount to the outer walls of the incubator and allows the user to set and adjust the shaker settings without opening the chamber door(s). A 6-position microplate platform is included and secures both standard or deep well plates.
Catalog No. 50-000-83115