Learn More
Chemglass Life Sciences OrbiShaker CO², Remote Controlled Orbital Shaker (for use with CLS-2450 45 & 180L Model Incubators)
SDP
Catalog No. 501979436
Encompass
OrbiShaker CO¿, Remote Controlled Orbital Shaker, for CLS-2450 45 & 180L Models. Orbi-Shaker CO2 series is designed for shaking, mixing & aeration applications in extreme environments, including CO2 incubators. It can withstand temperatures from O to 65¿C and humidity levels up to 100%. To avoid contamination, unintentional release of gas or deviations from protocol, all settings are adjusted via the included remote-control module. Supplied with a non-slip rubber mat platform. In addition, convenient MAGic Clamp TM platforms (of multiple sizes) are also available for use with Erlenmeyer flasks and test tube racks.
Catalog No. 50-197-9436 Supplier Chemglass Life Sciences Supplier No. CLS2450247
OrbiShaker CO¿, Remote Controlled Orbital Shaker, for CLS-2450 45 & 180L Models. Orbi-Shaker CO2 series is designed for shaking, mixing & aeration applications in extreme environments, including CO2 incubators. It can withstand temperatures from O to 65¿C and humidity levels up to 100%. To avoid contamination, unintentional release of gas or deviations from protocol, all settings are adjusted via the included remote-control module. Supplied with a non-slip rubber mat platform. In addition, convenient MAGic Clamp TM platforms (of multiple sizes) are also available for use with Erlenmeyer flasks and test tube racks.