Learn More
MSC Clarke Orbital Floor Machine, 1.5 hp, 3450 RPM, 20x14 in
Catalog No. 502838835
Encompass
This multipurpose electric floor machine features a 1.5 hp motor and 3450 RPM brush speed, designed for efficient cleaning. It has a 20 x 14 in cleaning width and is constructed with a durable aluminum housing, making it suitable for various floor maintenance tasks.
  • Multipurpose electric floor machine
  • 1.5 hp motor for powerful operation
  • 3450 RPM brush speed for effective cleaning
  • 20 x 14 in cleaning width
  • Durable aluminum housing
  • Equipped with a deadman switch for safety
  • 50 ft cord length
  • 13 A amperage
Catalog No. 50-283-8835 Supplier MSC Supplier No. 35706373
Add to Cart
Add to Cart
Request a Quote
This multipurpose electric floor machine features a 1.5 hp motor and 3450 RPM brush speed, designed for efficient cleaning. It has a 20 x 14 in cleaning width and is constructed with a durable aluminum housing, making it suitable for various floor maintenance tasks.
  • Multipurpose electric floor machine
  • 1.5 hp motor for powerful operation
  • 3450 RPM brush speed for effective cleaning
  • 20 x 14 in cleaning width
  • Durable aluminum housing
  • Equipped with a deadman switch for safety
  • 50 ft cord length
  • 13 A amperage