Learn More
MSC Clarke Focus II Boost Orbital Rider Scrubber, 28 in Cleaning Path, Orbital, AGM Battery Powered
Catalog No. 502838735
Encompass
The Focus II Boost Orbital Rider Scrubber is an AGM battery-powered floor scrubber with a 28 in cleaning path. It features a 31 gal solution and recovery tank, offering high capacity, long run times, and simple operation for various floor cleaning needs.
  • High capacity, long run times, simple operation
  • Equipped with conventional disc, cylindrical, or BOOST Technology scrub deck
  • Quiet operation for noise-sensitive areas
  • Optional chemical mixing system
  • Patented pulse-modulated solution control
  • Gimble-mounted brushes navigate uneven floors
  • Ergonomic controls
  • Onboard battery charger
  • 28 in cleaning width
  • 31 gal solution tank capacity
  • 31 gal recovery tank capacity
  • Polyethylene housing
  • 32.6094 in squeegee width
  • Deadman switch
  • 46 in water lift
  • 1.05 horsepower
  • Self-propelled
  • 2250 brush speed
Catalog No. 50-283-8735 Supplier MSC Supplier No. 35706613
Add to Cart
Add to Cart
Request a Quote
The Focus II Boost Orbital Rider Scrubber is an AGM battery-powered floor scrubber with a 28 in cleaning path. It features a 31 gal solution and recovery tank, offering high capacity, long run times, and simple operation for various floor cleaning needs.
  • High capacity, long run times, simple operation
  • Equipped with conventional disc, cylindrical, or BOOST Technology scrub deck
  • Quiet operation for noise-sensitive areas
  • Optional chemical mixing system
  • Patented pulse-modulated solution control
  • Gimble-mounted brushes navigate uneven floors
  • Ergonomic controls
  • Onboard battery charger
  • 28 in cleaning width
  • 31 gal solution tank capacity
  • 31 gal recovery tank capacity
  • Polyethylene housing
  • 32.6094 in squeegee width
  • Deadman switch
  • 46 in water lift
  • 1.05 horsepower
  • Self-propelled
  • 2250 brush speed