Learn More
MSC Clarke Floor Scrubber, 32 in Cleaning Width, Battery Powered, Orbital
Catalog No. 502838877
Encompass
The Clarke Focus II Boost Orbital Walk Behind Scrubber is a robust floor cleaning machine for optimal performance in commercial and industrial settings. With a 32 in cleaning width and battery power, this scrubber uses orbital technology for thorough and efficient floor cleaning, tackling challenging dirt and grime. Designed for ease of use and consistent results.
  • Versatile floor scrubber for various operations.
  • Battery powered for cordless operation and mobility.
  • 32 in cleaning width for broad coverage.
  • 23 gal recovery tank capacity.
  • 23 gal solution tank capacity.
  • 41 in squeegee width for effective water pickup.
  • 0.75 hp motor.
  • Durable polyethylene housing material.
  • Brush speed of 2250 RPM.
  • Equipped with a deadman switch for operator safety.
  • Self-propelled functionality for effortless operation.
Catalog No. 50-283-8877 Supplier MSC Supplier No. 35706928
Add to Cart
Add to Cart
Request a Quote
The Clarke Focus II Boost Orbital Walk Behind Scrubber is a robust floor cleaning machine for optimal performance in commercial and industrial settings. With a 32 in cleaning width and battery power, this scrubber uses orbital technology for thorough and efficient floor cleaning, tackling challenging dirt and grime. Designed for ease of use and consistent results.
  • Versatile floor scrubber for various operations.
  • Battery powered for cordless operation and mobility.
  • 32 in cleaning width for broad coverage.
  • 23 gal recovery tank capacity.
  • 23 gal solution tank capacity.
  • 41 in squeegee width for effective water pickup.
  • 0.75 hp motor.
  • Durable polyethylene housing material.
  • Brush speed of 2250 RPM.
  • Equipped with a deadman switch for operator safety.
  • Self-propelled functionality for effortless operation.