Learn More
MSC Clarke Floor Scrubber, 28 in Cleaning Path, AGM Battery Powered, Orbital
Catalog No. 502838733
Encompass
This mid-size scrubber is a reliable floor cleaning machine designed for efficiency. It offers various scrub deck options, including conventional disc, cylindrical for rough floors, and green-cleaning BOOST Technology, ensuring suitability for diverse scrubbing needs.
  • Variety of models for specific scrubbing needs.
  • Quiet operation for noise-sensitive areas.
  • Optional chemical mixing system ensures precise chemical-water mixture.
  • Gimble-mounted brushes navigate uneven floors for even scrubbing agitation.
  • Onboard battery charger allows recharging anywhere, reducing downtime.
Catalog No. 50-283-8733 Supplier MSC Supplier No. 35706969
Add to Cart
Add to Cart
Request a Quote
This mid-size scrubber is a reliable floor cleaning machine designed for efficiency. It offers various scrub deck options, including conventional disc, cylindrical for rough floors, and green-cleaning BOOST Technology, ensuring suitability for diverse scrubbing needs.
  • Variety of models for specific scrubbing needs.
  • Quiet operation for noise-sensitive areas.
  • Optional chemical mixing system ensures precise chemical-water mixture.
  • Gimble-mounted brushes navigate uneven floors for even scrubbing agitation.
  • Onboard battery charger allows recharging anywhere, reducing downtime.