Learn More
MSC Clarke Floor Scrubber, 32 in Cleaning Path, Battery Powered, 23 gal Tank, Orbital
Catalog No. 502839180
Encompass
This walk-behind floor scrubber is designed for efficient cleaning and maintaining spotless establishments. It operates quietly at 63 dB and features a 32 in cleaning path for maximum efficiency. The unit includes an adjustable chemical mixing system and is battery-operated for flexibility in cleaning large areas. BOOST technology enables chemical-free floor finish removal and provides high-speed agitation for superior cleaning on various hard floor types.
  • 32 in cleaning path
  • 23 gal solution and recovery tanks
  • Cordless, battery-powered operation
  • BOOST technology for chemical-free surface prep
  • Brush pressure adjustable from 95 to 150 lb
  • 2,250 RPM brush speed
  • 41 in squeegee length
  • Quiet 63 dB operation
  • Intuitive one-touch control panel
  • Adjustable chemical mixing system
  • Security key switch and thermal overload protection
Catalog No. 50-283-9180 Supplier MSC Supplier No. 35706936
Add to Cart
Add to Cart
Request a Quote
This walk-behind floor scrubber is designed for efficient cleaning and maintaining spotless establishments. It operates quietly at 63 dB and features a 32 in cleaning path for maximum efficiency. The unit includes an adjustable chemical mixing system and is battery-operated for flexibility in cleaning large areas. BOOST technology enables chemical-free floor finish removal and provides high-speed agitation for superior cleaning on various hard floor types.
  • 32 in cleaning path
  • 23 gal solution and recovery tanks
  • Cordless, battery-powered operation
  • BOOST technology for chemical-free surface prep
  • Brush pressure adjustable from 95 to 150 lb
  • 2,250 RPM brush speed
  • 41 in squeegee length
  • Quiet 63 dB operation
  • Intuitive one-touch control panel
  • Adjustable chemical mixing system
  • Security key switch and thermal overload protection