Learn More
Labnet International, Inc Orbit 1000 Digital Shaker, without Platform
Catalog No. NC0416500
Encompass_Preferred
Four platform configurations allow the Orbit 1000 Digital Orbital Shaker to meet a variety of shaking and mixing applications. Single and double flat platforms hold trays, plates and dishes. The double platform allows for expansion of the shaker capacity without increasing the footprint. The universal platform has spring loaded bars that adjust to fit flasks, bottles, tube racks and odd shaped containers. Pre-drilled to hold Erlenmeyer clamps (clamps sold separately), the flask platform is ideal for mixing and growing cultures. Platforms are removable and interchangeable.
Catalog No. NC0416500 Supplier Labnet International, Inc Supplier No. S20301000B
Four platform configurations allow the Orbit 1000 Digital Orbital Shaker to meet a variety of shaking and mixing applications. Single and double flat platforms hold trays, plates and dishes. The double platform allows for expansion of the shaker capacity without increasing the footprint. The universal platform has spring loaded bars that adjust to fit flasks, bottles, tube racks and odd shaped containers. Pre-drilled to hold Erlenmeyer clamps (clamps sold separately), the flask platform is ideal for mixing and growing cultures. Platforms are removable and interchangeable.