Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Made from medical-grade stainless steel for long-lasting durability and corrosion resistance
- Designed for precise eyelid retraction, maintaining optimal access to the orbital region
- Smooth, low-profile frame enhances comfort and reduces tissue irritation
- Balanced and ergonomic design supports surgeon control and reduces fatigue
- Autoclavable and reusable for reliable performance in sterile environments
- Provides a clear, unobstructed field for delicate orbital manipulation and dissection
- Cosmetic eyelid and reconstruction work
- Research and training involving ocular anatomy
Specifications
Specifications
| Product Type | Alfonso Eye Speculum |
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO, GMP |
| Color | Silver |
| Material | Stainless Steel |
| Grade | Surgical |
| Size | Standard |
| Sterility | Non-sterile |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?