Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Classic Barraquer-style eye speculum for reliable eyelid retraction
- Ideal for ophthalmic surgery, orbital dissection, and microsurgical work
- Thin, low-profile wire blades provide clear access to the operative field
- Smooth, contoured design minimizes pressure on delicate periocular tissue
- Lightweight construction improves surgeon control and patient comfort (or cadaver handling in educational labs)
- Crafted from surgical-grade stainless steel for durability and corrosion resistance
- Fully autoclavable and reusable
- Suitable for ophthalmology, oculoplastic procedures, research, and training environments
Specifications
Specifications
| Product Type | Barraquer Eye Speculum |
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO, GMP |
| Color | Silver |
| Material | Stainless Steel |
| Grade | Surgical |
| Size | Standard |
| Sterility | Non-sterile |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?