Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Built from medical-grade stainless steel for durability and corrosion resistance
- Provides firm yet gentle eyelid retraction to keep the surgical field unobstructed
- Stable frame design helps maintain consistent exposure throughout the procedure
- Smooth, contoured edges minimize tissue pressure and irritation
- Lightweight and ergonomic for precise handling during delicate orbital manipulation
- Autoclavable and reusable
- Orbital dissection and exploration
- Oculoplastic and reconstructive procedures
- Research and anatomical training settings
Specifications
Specifications
| Product Type | Knapp-Clark Eye Speculum |
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO, GMP |
| Color | Silver |
| Material | Stainless Steel |
| Grade | Surgical |
| Size | Standard |
| Sterility | Non-sterile |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?