Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Constructed from medical-grade stainless steel for durability and corrosion resistance
- Designed to gently retract eyelids while maintaining a stable surgical field
- Provides unobstructed access to the orbit for precise dissection and ophthalmic manipulation
- Smooth, contoured design enhances patient comfort and minimizes tissue trauma
- Lightweight and balanced for comfortable handling during extended procedures
- Autoclavable and reusable
- Ideal for environments requiring precise access and controlled retraction
- Orbital dissection and exploration
- Oculoplastic and cosmetic eyelid procedures
- Teaching and research environments involving eye anatomy
Specifications
Specifications
| Product Type | Lancaster Eye Speculum |
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO, GMP |
| Color | Silver |
| Material | Stainless Steel |
| Grade | Surgical |
| Size | Standard |
| Sterility | Non-sterile |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?