Learn More
Research Products International Corp Orbital-Genie, Benchtop, 18x18
SDP
Catalog No. 501976825
Encompass
A powerful large capacity orbital shaker designed to hold any type of container. Available in three platform versions.The fully programmable orbital shaker features variable speed including orbit reverse, timer from 1 minutes to 99 hours with alarm, programmable acceleration and deceleration rates. The ramping features ensures a smooth ramp-up to desired speed avoided potential spills. Operator can create any mixing or shaking action from a smooth vortex to sloshing aeration (or anything in between). The unit can be used in cold room or incubator and CO2 environments. Available in three platform options as follows:
  • Orbital-Genie with Universal Ratcheting Clamps – provides the flexibility for using a mix of different size and shape vessels on a single platform. Unit is supplied with two ratcheting cushioned clamps. Extra ratcheting clamps for more than two rows of smaller vessels are optional.
Catalog No. 50-197-6825 Supplier Research Products International Corp Supplier No. 155601
A powerful large capacity orbital shaker designed to hold any type of container. Available in three platform versions.The fully programmable orbital shaker features variable speed including orbit reverse, timer from 1 minutes to 99 hours with alarm, programmable acceleration and deceleration rates. The ramping features ensures a smooth ramp-up to desired speed avoided potential spills. Operator can create any mixing or shaking action from a smooth vortex to sloshing aeration (or anything in between). The unit can be used in cold room or incubator and CO2 environments. Available in three platform options as follows:
  • Orbital-Genie with Universal Ratcheting Clamps – provides the flexibility for using a mix of different size and shape vessels on a single platform. Unit is supplied with two ratcheting cushioned clamps. Extra ratcheting clamps for more than two rows of smaller vessels are optional.