Learn More
Bellco Glass, Inc. Orbital Shaker,12V/115V With 25x25cm Tray
SDP
Catalog No. 501946219
Encompass_Preferred
The Bellco btbTM12 Volt Orbital Shaker is a compact unit designed for continuous duty shaking using intrinsically safe low voltage electronics. The one-inch orbital motion provides even mixing. The rubber suction cup feet keep the unit firmly attached to surface. Counterbalanced drive shafts for precise orbital motion. Includes 25x25cm (10x10" ) tray. Dims: 11x11x5" (28x28x14cm). NOTE: CAN USE THE UNIVERSAL TRAYS WITH CLAMPS BUT ONLY THE SMALL TRAYS - 25X25. MADE IN THE USA
Catalog No. 50-194-6219 Supplier Bellco Glass, Inc. Supplier No. 777730110
The Bellco btbTM12 Volt Orbital Shaker is a compact unit designed for continuous duty shaking using intrinsically safe low voltage electronics. The one-inch orbital motion provides even mixing. The rubber suction cup feet keep the unit firmly attached to surface. Counterbalanced drive shafts for precise orbital motion. Includes 25x25cm (10x10" ) tray. Dims: 11x11x5" (28x28x14cm). NOTE: CAN USE THE UNIVERSAL TRAYS WITH CLAMPS BUT ONLY THE SMALL TRAYS - 25X25. MADE IN THE USA