Learn More
JDM LAB SOLUTIONS LLC Orbital Shaker / Designed for CO2 Incubators
Supplier:  JDM LAB SOLUTIONS LLC OSC26M
Encompass
Remote Control attaches externally to CO2IncubatorLED display shows both set and actual parameters without opening the doorSimple and intuitive design provides easy operationAlarm function alerts the user when the timer reaches zeroShaker is easy to remove for cleaning/disinfecting to protect your cellsThe Magnetic Orbital Drive system uses magnetic force to deliver precise speed control, low vibration, maintenance free operation and long service lifeSuitable for use in CO2, Standard or Refrigerated Incubators at up to 20% CO2 and 90% Humidity environmentsWide range of accessories to accommodate various sample holdersDeveloped to help you economically scale-up cell production in your existing CO2 Incubator. Designed with sealed electronics to minimize heat disruption and ensure that conditions in the CO2 chamber are not affected.
Catalog No. NC2385862