Learn More
Research Products International Corp Scienceware Orbital Shaker, Rotating
SDP
Catalog No. 50999286
Encompass
Turns a standard magnetic stirrer into an orbital shaking platform saving the expense and space requirements of additional lab equipment. Perfect for dishes, flasks, and trays, it is also ideal for Western Blot processing.
Shake and mix; rotating orbit diameter of 21.7 mm (7/8 inch)
Shaker adjustment is controlled by setting the magnetic stirrer to the desired speed
Platform protects the operating mechanism from spills; non-skid surface prevents containers from sliding
Base incorporates built-in hand holds for lifting and carrying
Supports up to 6.8 kg (15 pounds)
Catalog No. 50-999-286 Supplier Research Products International Corp Supplier No. 437040
Turns a standard magnetic stirrer into an orbital shaking platform saving the expense and space requirements of additional lab equipment. Perfect for dishes, flasks, and trays, it is also ideal for Western Blot processing.
Shake and mix; rotating orbit diameter of 21.7 mm (7/8 inch)
Shaker adjustment is controlled by setting the magnetic stirrer to the desired speed
Platform protects the operating mechanism from spills; non-skid surface prevents containers from sliding
Base incorporates built-in hand holds for lifting and carrying
Supports up to 6.8 kg (15 pounds)