Learn More
GRANT INSTRUMENTS Orbital shaker multi-platform. Order platform separately
SDP
Catalog No. NC1476974
Encompass_Preferred
Orbital platform shaker PSU-10i is a benchtop multifunctional shaking unit.Microprocessor controlled shaking platform providing smooth and quiet horizontal orbital motion for mixing in bottles, flasks and beakers.Variable shaking speed: 50 to 450 rpm.Interchangeable platforms for use with a variety of vessels.Loads up to 3 kg.Continuous or timed operation with automatic switch-off.Reliable direct drive system.Automatic load balancing system.Soft start feature ensures a smooth ramp-up - avoids potential damage to vessel content.Exceptionally quiet motor - consistent even shaking.
Catalog No. NC1476974 Supplier GRANT INSTRUMENTS Supplier No. PSU10I
Orbital platform shaker PSU-10i is a benchtop multifunctional shaking unit.Microprocessor controlled shaking platform providing smooth and quiet horizontal orbital motion for mixing in bottles, flasks and beakers.Variable shaking speed: 50 to 450 rpm.Interchangeable platforms for use with a variety of vessels.Loads up to 3 kg.Continuous or timed operation with automatic switch-off.Reliable direct drive system.Automatic load balancing system.Soft start feature ensures a smooth ramp-up - avoids potential damage to vessel content.Exceptionally quiet motor - consistent even shaking.