Learn More
Cole-Parmer SI-200D-120 Stuart Shaking Incubator, 51 L; 120 VAC
Catalog No. NC1490811 Shop All Cole-Parmer Products
Encompass
These shaking incubators provide an orbital shaking motion to ensure even mixing while promoting bacterial growth. All models feature a timer which can be set from 1 second to 9 days. Once the timer has counted down, an alarm will sound and the shaking action will stop while the incubator will continue to run.The retractable platform allows easy access to the samples at the back of the incubator yet securely locks in place during operation. A versatile clamping system holds flasks securely in place (up to 1 L flasks for 51 L models and up to 2 L flasks for 115 L models). Built-in USB connection and dedicated software allow long-term monitoring of the incubator temperature via a PC.Please Note: Effective May 2022, the Stuart Shaking Incubator Model number SI500/120/60 has now been rebranded Cole-Parmer under Model number SI-200D-120. All the same great features and specifications, just with a new and improved look.
Catalog No. NC1490811 Supplier Cole-Parmer Supplier No. 7952000
These shaking incubators provide an orbital shaking motion to ensure even mixing while promoting bacterial growth. All models feature a timer which can be set from 1 second to 9 days. Once the timer has counted down, an alarm will sound and the shaking action will stop while the incubator will continue to run.The retractable platform allows easy access to the samples at the back of the incubator yet securely locks in place during operation. A versatile clamping system holds flasks securely in place (up to 1 L flasks for 51 L models and up to 2 L flasks for 115 L models). Built-in USB connection and dedicated software allow long-term monitoring of the incubator temperature via a PC.Please Note: Effective May 2022, the Stuart Shaking Incubator Model number SI500/120/60 has now been rebranded Cole-Parmer under Model number SI-200D-120. All the same great features and specifications, just with a new and improved look.