Learn More
Research Products International Corp Orbitron Rotator, Model II
SDP
Catalog No. 50212679
Encompass
Orbitron II Rotator provides a gentle mixing action for low foaming applications. Ideal for staining, destaining and blotting procedures. Compact design allows use in most incubators and saves valuable bench space. Speed adjusts from 6 to 23 orbits per minute. Platform angle has a 0° to 23° adjustable tilt.
Supplied with platform, one flat non-skid mat, one dimpled mat for tubes, low voltage power supply and two year warranty.
Catalog No. 50-212-679 Supplier Research Products International Corp Supplier No. 260250
Orbitron II Rotator provides a gentle mixing action for low foaming applications. Ideal for staining, destaining and blotting procedures. Compact design allows use in most incubators and saves valuable bench space. Speed adjusts from 6 to 23 orbits per minute. Platform angle has a 0° to 23° adjustable tilt.
Supplied with platform, one flat non-skid mat, one dimpled mat for tubes, low voltage power supply and two year warranty.