Learn More
MilliporeSigma™ Supelco™ ORBO™ 2000 Precleaned Large PUF Plug, Replacement
Catalog No. 111004852 Shop All MilliporeSigma Supelco Products
Click to view available options
Diameter (Metric):
6 cm
Used for gas and vapor sampling
PUF plugs of appropriate density effectively trap contaminants without creating excessive back pressure. PUF plugs sometimes are used with an additional adsorbent, such as Amberlite™ XAD™-2 resin, to collect more volatile contaminants, or with a filter, to trap particles.
TRUSTED_SUSTAINABILITY
Specifications
Description Replacement
Quantity 1/Pk.
Length (Metric) 7.6 cm
Diameter (Metric) 6 cm
ORBO is a trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.