Learn More
MilliporeSigma™ Supelco™ ORBO™ 351 4-TBC on Activated Coconut Charcoal (20/40), 100/50 mg
Shop All MilliporeSigma Supelco ProductsDescription
- Contains two beds of the same selective adsorbent (Bed A: 100 mg, Bed B: 50 mg) separated by glass wool or foam
- Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed
- Matrix: 4-tert-butyl catechol on Charcoal
Specifications
Specifications
| Description | 4-tert-butyl catechol on charcoal |
| For Use With (Application) | Solvent Desorption |
| Quantity | 50 |
| Type | Solvent Desorption Tube |
| Length (Metric) | 75 mm |
| Diameter (Metric) | 6 mm |
ORBO is a trademark of Sigma-Aldrich Co. LLC
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.