Learn More
MilliporeSigma™ Supelco™ ORBO™ 65P (OVS) Amberlite XAD-4 specially cleaned, 160/80 mg RC,GFF,F,F (specially cleaned) separators
Catalog No. 111009878 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
13-8 mm
Catalog No. Diameter (Metric)
11-100-9878 13-8 mm
1 options
Catalog No. 11-100-9878 Supplier MilliporeSigma™ Supelco™ Supplier No. 20029U
Only null left
Add to Cart
Add to Cart
Used for gas and vapor sampling
ORBO™ sorbent tubes contain two beds of the same selective adsorbent separated by glass wool or foam, for gas and vapor sampling. The dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
TRUSTED_SUSTAINABILITY
Specifications
Description Bed A 160 mg/Bed B 80 mg
Material RC,GFF,F,F (Specially Cleaned) Separators
Quantity 10/Pk.
Length (Metric) 75 mm
Diameter (Metric) 13-8 mm
ORBO is a trademark of Sigma-Aldrich Co. LLC