Learn More
MilliporeSigma™ Supelco™ ORBO™ 78 HBr on Carboxen 564 specially cleaned (20/45), 400/200 mg W,W,W separators
Catalog No. 111004568 Shop All MilliporeSigma Supelco Products
Click to view available options
Length (Metric):
110 mm
Used for gas and vapor sampling
ORBO™ 78 HBR on Carboxen™ 564 specially cleaned (20/45), 400/200 mg sorbent tubes (W,W,W separators, O.D. x L 6 mm x 110 mm) contain two beds of the same selective adsorbent separated by glass wool or foam, for gas and vapor sampling. The dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
TRUSTED_SUSTAINABILITY
Specifications
Material W,W,W Separators
Quantity 25/Pk.
Length (Metric) 110 mm
Diameter (Metric) 6 mm
ORBO is a trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.