Learn More
MilliporeSigma™ Supelco™ ORBO™ 90 Carboxen 564 (20/45), 160/80 mg W,F,F separators
Catalog No. 111004703 Shop All MilliporeSigma Supelco Products
Click to view available options
Length (Metric):
75 mm
Used for gas and vapor sampling
ORBO™ 90 Carboxen™ 564 (20/45), 160/80 mg solvent desorption tube is designed for gas and vapor sampling. It contains two beds of the same selective adsorbent separated by glass wool or foam. Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.
TRUSTED_SUSTAINABILITY
Specifications
Material W,F,F Separators
Quantity 25/Pk.
Length (Metric) 75 mm
Diameter (Metric) 6 mm
ORBO is a trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.