Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ ORBO-DNPH LpDNPH on Silica Gel, 120 mg PE separators

Catalog No. 111009676 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
90 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-100-9676 90 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-9676 Supplier MilliporeSigma™ Supelco™ Supplier No. 20081U
Add to Cart
Add to Cart

Used for gas and vapor sampling

The LpDNPH ORBO™ Tube with frangible seal is designed for determination of low-level carbonyl (e.g formaldehyde) emissions from building materials. The sorbent tube is 1/4 in. O.D. x 90 mm long with frangible design at one end. It contains 120 mg of DNPH coated silica gel and is capped with PTFE caps.
TRUSTED_SUSTAINABILITY

Specifications

Material PE Separators
Quantity 10/Pk.
Length (Metric) 90 mm
Diameter (Metric) 6 mm

ORBO is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.