Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Orbofiban acetate | 163250-91-7 | 98.1% | 421.45 | C19H27N5O6 | 25 MG
SDP

Catalog No. 5000246145
Encompass_Preferred

Orbofiban acetate is an orally active platelet GPIIb/IIIa antagonist that inhibits platelet aggregation. It is supplied for research use only and is not for human or clinical use.

  • Potent platelet GPIIb/IIIa antagonist that inhibits platelet aggregation.
  • Purity 98.13%.
  • Molecular formula C19H27N5O6 and molecular weight 421.45 g/mol.
  • Available as 25 mg vials for small-scale studies.
  • Soluble in DMSO: 150 mg/mL (sonication and warming to 60°C may be required; use newly opened DMSO).
  • Intended for in vitro and in vivo research; not for human use.

Catalog No. 50-002-46145 Supplier Medchemexpress LLC Supplier No. HY10304A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Orbofiban acetate is an orally active platelet GPIIb/IIIa antagonist that inhibits platelet aggregation. It is supplied for research use only and is not for human or clinical use.

  • Potent platelet GPIIb/IIIa antagonist that inhibits platelet aggregation.
  • Purity 98.13%.
  • Molecular formula C19H27N5O6 and molecular weight 421.45 g/mol.
  • Available as 25 mg vials for small-scale studies.
  • Soluble in DMSO: 150 mg/mL (sonication and warming to 60°C may be required; use newly opened DMSO).
  • Intended for in vitro and in vivo research; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.