Learn More
Medchemexpress LLC Orbofiban acetate | 163250-91-7 | 98.1% | 421.45 g/mol | C19H27N5O6 | 1 ML
SDP
Catalog No. 5000247497
Encompass_Preferred
Orbofiban acetate is an orally active platelet GPIIb/IIIa antagonist that inhibits platelet aggregation and is supplied for research use only. It is available as a 10 mM solution in DMSO (1 mL) or as a solid in multiple milligram sizes.
  • Orally active GPIIb/IIIa antagonist
  • Inhibits platelet aggregation
  • Available as 10 mM solution in DMSO (1 mL) and as solid in 5-100 mg sizes
  • High purity (98.13%)
  • Storage: solid at 4°C under nitrogen; in solvent at -80°C (up to 6 months) or -20°C (up to 1 month)
  • Intended for research use only
Catalog No. 50-002-47497 Supplier Medchemexpress LLC Supplier No. HY10304A10MM 1ML
Orbofiban acetate is an orally active platelet GPIIb/IIIa antagonist that inhibits platelet aggregation and is supplied for research use only. It is available as a 10 mM solution in DMSO (1 mL) or as a solid in multiple milligram sizes.
  • Orally active GPIIb/IIIa antagonist
  • Inhibits platelet aggregation
  • Available as 10 mM solution in DMSO (1 mL) and as solid in 5-100 mg sizes
  • High purity (98.13%)
  • Storage: solid at 4°C under nitrogen; in solvent at -80°C (up to 6 months) or -20°C (up to 1 month)
  • Intended for research use only