Learn More
Medchemexpress LLC Orbofiban acetate | 163250-91-7 | 98.1% | 421.45 g/mol | C19H27N5O6 | 10 MG
SDP
Catalog No. 5000247465
Encompass_Preferred
Orbofiban acetate is an orally active platelet GPIIb/IIIa antagonist used in research to inhibit platelet aggregation. It is supplied as the acetate salt for laboratory research use only and is not for human or clinical use.
  • Orally active GPIIb/IIIa antagonist that inhibits platelet aggregation.
  • Supplied as acetate salt with reported purity of 98.13%.
  • Available in multiple small research package sizes (5-500 mg) suitable for in vitro and in vivo studies.
  • Soluble in DMSO, facilitating preparation of concentrated stock solutions for assays.
  • Used to study platelet aggregation, thrombosis models, and GPIIb/IIIa receptor function.
Catalog No. 50-002-47465 Supplier Medchemexpress LLC Supplier No. HY10304A10MG
Orbofiban acetate is an orally active platelet GPIIb/IIIa antagonist used in research to inhibit platelet aggregation. It is supplied as the acetate salt for laboratory research use only and is not for human or clinical use.
  • Orally active GPIIb/IIIa antagonist that inhibits platelet aggregation.
  • Supplied as acetate salt with reported purity of 98.13%.
  • Available in multiple small research package sizes (5-500 mg) suitable for in vitro and in vivo studies.
  • Soluble in DMSO, facilitating preparation of concentrated stock solutions for assays.
  • Used to study platelet aggregation, thrombosis models, and GPIIb/IIIa receptor function.