Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Orbofiban acetate | 163250-91-7 | 98.1% | 421.45 g/mol | C19H27N5O6 | 100 MG
SDP

Catalog No. 5000247004
Encompass_Preferred

Orbofiban acetate is an orally active platelet GPIIb/IIIa antagonist used in research to inhibit platelet aggregation. It is provided as a solid or as a DMSO solution and intended for in vitro and preclinical studies.

  • Orally active platelet GPIIb/IIIa antagonist that inhibits platelet aggregation.
  • High purity (approximately 98.1%).
  • Molecular formula C19H27N5O6; molecular weight 421.45 g/mol.
  • Available as solid (5-500 mg) and as a 10 mM solution in DMSO.
  • Soluble in DMSO (≈150 mg/mL) with warming and sonication.
  • Storage: solid at 4°C under inert atmosphere; solutions at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-47004 Supplier Medchemexpress LLC Supplier No. HY10304A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Orbofiban acetate is an orally active platelet GPIIb/IIIa antagonist used in research to inhibit platelet aggregation. It is provided as a solid or as a DMSO solution and intended for in vitro and preclinical studies.

  • Orally active platelet GPIIb/IIIa antagonist that inhibits platelet aggregation.
  • High purity (approximately 98.1%).
  • Molecular formula C19H27N5O6; molecular weight 421.45 g/mol.
  • Available as solid (5-500 mg) and as a 10 mM solution in DMSO.
  • Soluble in DMSO (≈150 mg/mL) with warming and sonication.
  • Storage: solid at 4°C under inert atmosphere; solutions at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.