Learn More
Cayman Chemical ORC1 BAH domain (human, recombinant), Purity: ≥80%, 1 mg
SDP
Catalog No. 502953181
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • MW: 48.2 kDa
Catalog No. 50-295-3181 Supplier Cayman Chemical Supplier No. 119191 MG
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • MW: 48.2 kDa