Learn More
Aviva Systems Biology Corp ORC3L Antibody - N-terminal region (ARP62224_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62224_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ORC3L. It was validated on Western Blot. . Alternate names: LAT, LATHEO, ORC3, ORC3LApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-619