Learn More
Aviva Systems Biology Corp ORC6L Antibody - C-terminal region (ARP55022_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55022_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ORC6L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ORC6L. Alternate names: ORC6, ORC6LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61398