Learn More
Medchemexpress LLC Orcein | 1400-62-0 | 1 G
SDP
Catalog No. 5000230633
Encompass_Preferred
Orcein is an irreversible histological stain used to visualize elastic fibers and related tissue components in biological specimens. It binds hydrophobically to protein components of elastic fibers, producing purple to purple-red staining, and is used for elastic fiber analysis and morphological studies.
  • Irreversible stain selective for elastic fibers.
  • Produces purple to purple-red coloration of stained tissues.
  • Useful for histology, atherosclerotic plaque analysis, and chromosome visualization.
  • Solid form; appearance reddish brown to black.
  • Store at 4°C and protect from light; stable in solvent at -80°C for 6 months.
Catalog No. 50-002-30633 Supplier Medchemexpress LLC Supplier No. HY1222661G
Orcein is an irreversible histological stain used to visualize elastic fibers and related tissue components in biological specimens. It binds hydrophobically to protein components of elastic fibers, producing purple to purple-red staining, and is used for elastic fiber analysis and morphological studies.
  • Irreversible stain selective for elastic fibers.
  • Produces purple to purple-red coloration of stained tissues.
  • Useful for histology, atherosclerotic plaque analysis, and chromosome visualization.
  • Solid form; appearance reddish brown to black.
  • Store at 4°C and protect from light; stable in solvent at -80°C for 6 months.