Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orcein Natural, Thermo Scientific™

Catalog No. AC151260025
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC151260025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC151260025 Supplier Thermo Scientific Chemicals Supplier No. 151260025

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1400-62-0
Molecular Formula C28H24N2O7
Molecular Weight (g/mol) 500.507
InChI Key VPEASJIRGSVXBF-UHFFFAOYSA-N
Synonym Natural Red 28
PubChem CID 5386447
IUPAC Name 2,8-bis(2,4-dihydroxy-6-methylanilino)-1,9-dimethyldibenzofuran-3,7-dione
SMILES CC1=CC(=CC(=C1NC2=C(C3=C4C(=C(C(=O)C=C4OC3=CC2=O)NC5=C(C=C(C=C5C)O)O)C)C)O)O

Specifications

Absorbance 200 to 400 (at lambda max.)
Beilstein 06, 885
Merck Index 15, 6958
Infrared Spectrum Authentic
Lambda Max 575 to 580nm (in 0.01M NaOH)
Formula Weight 500.51
Loss on Drying 5% max. (110°C)
Color Black/Brown to Brown
Quantity 2.5 g
Physical Form Powder
Chemical Name or Material Orcein, natural
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.