Learn More
Bioworld Orchid Maintenance Medium, w/ Charcoal and Agar, 50 L
Catalog No. 50254967
Encompass_Preferred
Orchid Maintenance MediumContains Micronutrients, Macronutrients, vitamins, sucrose, peptone and MES buffer required for orchid maintenance. Also contains Charcoal and AgarAdd auxins and cytokinins as needed.Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 35.31 g/LpH: 4.75–5.75Partially soluble in water
Catalog No. 50-254-967 Supplier Bioworld Supplier No. 306301884
Orchid Maintenance MediumContains Micronutrients, Macronutrients, vitamins, sucrose, peptone and MES buffer required for orchid maintenance. Also contains Charcoal and AgarAdd auxins and cytokinins as needed.Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 35.31 g/LpH: 4.75–5.75Partially soluble in water