Learn More
Bioworld Orchid Maintenance/ Replate Medium, w/ Banana and Charcoal, 50 L
Catalog No. 50254970
Encompass_Preferred
Orchid Maintenance/ Replate MediumContains Micronutrients and Macronutrients required to maintain/ replate orchidsContains Banana and CharcoalSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 57.31 g/LpH: 4.5–5.5Partially soluble in water
Catalog No. 50-254-970 Supplier Bioworld Supplier No. 306301894
Orchid Maintenance/ Replate MediumContains Micronutrients and Macronutrients required to maintain/ replate orchidsContains Banana and CharcoalSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 57.31 g/LpH: 4.5–5.5Partially soluble in water