Learn More
Bioworld Orchid Multiplication Medium, 50 L
Catalog No. 50254976
Encompass_Preferred
Orchid Multiplication MediumContains Micronutrients and Macronutrients required for orchid multiplicationSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 32.30 g/LpH: 4.75–5.75Partially soluble in water
Catalog No. 50-254-976 Supplier Bioworld Supplier No. 306301914
Orchid Multiplication MediumContains Micronutrients and Macronutrients required for orchid multiplicationSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 32.30 g/LpH: 4.75–5.75Partially soluble in water