Learn More
Bioworld Orchid Seed Sowing Medium, w/ Agar, 50 L
Catalog No. 50254982
Encompass_Preferred
Orchid Seed Sowing MediumContains Micronutrients and Macronutrients required for orchid seed sowingContains AgarSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 31.74 g/LPartially soluble in water
Catalog No. 50-254-982 Supplier Bioworld Supplier No. 306301934
Orchid Seed Sowing MediumContains Micronutrients and Macronutrients required for orchid seed sowingContains AgarSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 31.74 g/LPartially soluble in water