Learn More
Bioworld Orchid Seed Sowing Medium, w/ Agar & Charcoal, 50 L
Catalog No. 50254979
Encompass_Preferred
Orchid Seed Sowing MediumContains Micronutrients and Macronutrients required for orchid seed sowingContains Charcoal and AgarSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 32.74 g/LpH: 5.25–6.25Partially soluble in water
Catalog No. 50-254-979 Supplier Bioworld Supplier No. 306301924
Orchid Seed Sowing MediumContains Micronutrients and Macronutrients required for orchid seed sowingContains Charcoal and AgarSucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 32.74 g/LpH: 5.25–6.25Partially soluble in water